Order Entry
Denmark
ContactUsLinkComponent
AQUAMAX 2K BUNDLE Includes: INSTRUMENT, AQUAMAX 2000 MICROPLATE WASHER (P/N 0310-5363, qty 1) HEAD, 96 PLATE WASH AQUAMAX 2000/400 0 (P/N 0310-5216T, qty 1) ASSY, BOTTLE, 4 LITER, WITH LEVEL SENSOR (P/N 5000802, qty 2) 10L WASTE RESERVOIR (SK 20462, 1 * 1 items
  MLDVAQMAX2KBNDLTR
AQUAMAX 2K BUNDLE Includes: INSTRUMENT, AQUAMAX 2000 MICROPLATE WASHER (P/N 0310-5363, qty 1) HEAD, 96 PLATE WASH AQUAMAX 2000/400 0 (P/N 0310-5216T, qty 1) ASSY, BOTTLE, 4 LITER, WITH LEVEL SENSOR (P/N 5000802, qty 2) 10L WASTE RESERVOIR (SK 20462, 1 * 1 items
  MLDVAQMAX2KBNDLTR
Frequently Bought Together