Order Entry
Denmark
ContactUsLinkComponent
The ScanLater Western Blot cartridge option, user-installable to any SpectraMax i3/i3X or SpectraMax Paradigm base system. The Scan Later Western Blot detection system includes the ScanLater Western Blot Detection Cartridge, ScanLater Membrane Holder 1 * 1 items
Catalog # MLDV0200-7027-TR
Supplier:  Molecular Devices
The ScanLater Western Blot cartridge option, user-installable to any SpectraMax i3/i3X or SpectraMax Paradigm base system. The Scan Later Western Blot detection system includes the ScanLater Western Blot Detection Cartridge, ScanLater Membrane Holder 1 * 1 items
Catalog # MLDV0200-7027-TR
Supplier:  Molecular Devices
Frequently Bought Together