Order Entry
Denmark
ContactUsLinkComponent
SpectraMax AlphaScreen 384 HTS Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScreen assays. Compatible with SpectraMax PARADIGM and SpectraMax i3x systems. In addition, a patent pending design that is 1 * 1 items
Catalog # MLDV0200-7018-TR
Supplier:  Molecular Devices
SpectraMax AlphaScreen 384 HTS Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScreen assays. Compatible with SpectraMax PARADIGM and SpectraMax i3x systems. In addition, a patent pending design that is 1 * 1 items
Catalog # MLDV0200-7018-TR
Supplier:  Molecular Devices
Frequently Bought Together