Bottles, narrow neck, with thread, SYSTEM PLUS® 32
160 ml, DIN 32
Catalog # TOUR6461S1
Supplier: TOURNAIRE-EMBALLAGE
Bottles, narrow neck, with thread, SYSTEM PLUS® 32
Catalog # TOUR6461S1
Supplier: TOURNAIRE-EMBALLAGE
Jump to:
Product Details & Documents
Pure aluminium.
- Unbreakable, odorless
- Very good temperature resistance
- Leakproof with tamper-evident screw caps (must be odered separately)
- UN-approved
Suitable for storage and transport of highly aggessive liquids.
Documents
Specifications
- Thread:DIN 32
- Capacity:160 ml
- Seq:06