Recombinant Human Plasma Kallikrein 1B Protein 1 * 10 µG
Catalog # ABCAAB307350-10
Supplier: Abcam
Frequently Bought Together
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
PCR tubes, 0.2 ml, with flat or domed cap, ABgene®
Supplier: Thermo Fisher Scientific
VWR®, Deep Well Plates, 96 Square Well, Reinforced for Genomics Applications
Supplier: VWR Collection
Ice pans and buckets, multi-purpose, Corning®
Supplier: Corning
Racks and lids for storage 2D barcoded tubes, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
AcroPrep™ 24-Well Receiver Plate
Supplier: Cytiva (Formerly Pall Lab)