Order Entry
Denmark
Orders LinkContactUsLinkComponent
Liquid Dip Samplers

Liquid Dip Samplers

Liquid Dip sampler, Stainless steel 316L, 100 ml
Catalog # SAMP5175A-100
Supplier:   Sampling Systems
Liquid Dip Samplers
Catalog # SAMP5175A-100
Supplier:  Sampling Systems
Liquid Dip Samplers
Liquid Dip Samplers

Liquid Dip sampler, Stainless steel 316L, 100 ml

Price
2.980,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

The stainless steel liquid dip sampler is used to collect a wide variety of liquid samples from the surface.

  • Made from high quality and chemically resistant 316L stainless steel
  • Different sizes available
  • Convenient hands-free emptying feature

Once the sample has been collected, the liquid dip sampler can simply be pushed against the container allowing for the liquid to be easily emptied.

Specifications
  • Capacity:
    100 ml
  • Description:
    Liquid Dip sampler
  • Material:
    Stainless steel 316L
  • Seq:
    02