VITAMIN B5, CALCIUM SALT:H2O (CALCIUM PANTOTHENATE:H2O)(BETA-ALANYL-13C3, 99% 15N, 98%) 1 * 10 mg
Catalog # CAMBCNLM-7694-0.01
Supplier: CAMBRIDGE ISOTOPE LABS
VITAMIN B5, CALCIUM SALT:H2O (CALCIUM PANTOTHENATE:H2O)(BETA-ALANYL-13C3, 99% 15N, 98%) 1 * 10 mg
Catalog # CAMBCNLM-7694-0.01
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
TETRASUL 1 * 50 mg
Supplier: Merck
Isoproturon certified 1 * 0,1 g
Supplier: EHRENSTORFER
Tepraloxydim certified 1 * 0,1 g
Supplier: EHRENSTORFER
1-Bromo-2-chloroethane
Supplier: EHRENSTORFER
Supelco® PESTANAL® Organic Reference Standard, Amitrol
Supplier: Merck
Gadolinium standard solution, 1000 mg/L Gd in 5% nitric acid (de Gd?O?), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Supelco® Organic Reference Standard, α-Arbutin
Supplier: Merck
n-Pentadecane, Supelco®
Supplier: Merck
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Hardcheck p/total hardness tablets in pack of 250 1 * 250 Tablet
Supplier: Lovibond
Buffer solutions
Supplier: REAGECON
Enrofloxacin, analytical standard, VETRANAL™, Supelco®
Supplier: Merck
Specpure® Tungsten Standard Solution, 1000 mg/L W in Water (from (NH₄)₂WO₄) Standard for ICP
Supplier: Thermo Fisher Scientific