Internal Standard Mix - 7 components: 10ug/ml each of 6Li Sc Ge Y 1 * 100 mL
Catalog # ACSDN9303832.L1
Supplier: CPACHEM
Internal Standard Mix - 7 components: 10ug/ml each of 6Li Sc Ge Y 1 * 100 mL
Catalog # ACSDN9303832.L1
Supplier: CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW