VIAL RACK BLUE POLYPROPYLENE FOR 12 X 32MM VIALS 50 CAVITIES (5 X 10) NO LID REUSABLE 20 X 10.5 X 2.2 CM 1/PACK 1 * 1 items
Catalog # HYPE6EVR1232B
Supplier: Thermo Fisher Scientific
VIAL RACK BLUE POLYPROPYLENE FOR 12 X 32MM VIALS 50 CAVITIES (5 X 10) NO LID REUSABLE 20 X 10.5 X 2.2 CM 1/PACK 1 * 1 items
Catalog # HYPE6EVR1232B
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
(±)-1,2-Pentanediol 96%
Supplier: Thermo Fisher Scientific