Cell Scraper, Pivoting Blade, 30 cm 1 * 100 items
Catalog # STMC200-0598
Supplier: STEMCELL Technologies
Cell Scraper, Pivoting Blade, 30 cm 1 * 100 items
Catalog # STMC200-0598
Supplier: STEMCELL Technologies
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Supplier: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Laboratory trays
Supplier: KARTELL
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Aspirating pipettes, Falcon®
Supplier: Corning
Cryogenic tubes
Supplier: Brand
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection