Order Entry
Denmark
ContactUsLinkComponent
CUSTOM PACKED COLUMN Hayesep D60/80, 0,8meter, 2,0 mmID, 1/8 in OD SilcoSmoth Tubing, OC, Precond, no kit, Inlet & Outlet packed full:NO Packed Column Special Instruction FIGURE 2 A=0.125IN, B=2,5CM, X &Y=4CM, S=2,5cm 1 * 1 Items
Catalog # RESTPKC57280
Supplier:  Restek
CUSTOM PACKED COLUMN Hayesep D60/80, 0,8meter, 2,0 mmID, 1/8 in OD SilcoSmoth Tubing, OC, Precond, no kit, Inlet & Outlet packed full:NO Packed Column Special Instruction FIGURE 2 A=0.125IN, B=2,5CM, X &Y=4CM, S=2,5cm 1 * 1 Items
Catalog # RESTPKC57280
Supplier:  Restek
Frequently Bought Together