50 TABLETS NITRITE HR 1 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSHNIHR150
Supplier: WATER ID PRIMELAB
50 TABLETS NITRITE HR 1 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSHNIHR150
Supplier: WATER ID PRIMELAB
Frequently Bought Together
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Ammonia solution ∼32%, GPR RECTAPUR®
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck
Test strips, peroxide, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals