50 POWDER-PILLOWS NITRATE HR N 2 1 * 50 items
Catalog # WTIDPPPNITRA250
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Test kits, hydrogen peroxide, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Kjeldahl tablets, Kjeltabs
Supplier: THOMPSON & CAPPER
Ethanol 70% ≥70% (v/v)
Supplier: VWR Chemicals
Supelco® Spectroquant® Test Kits, Sulphate
Supplier: Merck
Schiff reagent for analysis of aldehydes
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Supplier: VWR Chemicals