REAGENT KIT FOR PRIMELAB ID14 (50 TESTS) 1 * 1 items
Catalog # WTIDPL14-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID14 (50 TESTS) 1 * 1 items
Catalog # WTIDPL14-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Urine test strips, Medi-Test
Supplier: MACHEREY-NAGEL
Phenolphthalein 0.5 wt. % in ethanol: water (1:1), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Bromocresol green-methyl red solution mixed indicator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Acid red 27 ≥85-95% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Vario Chlorine Free DPD F10, Powder Packs in Bag of 100 1 * 100 items
Supplier: Lovibond
NANOCOLOR® Photometric Tube Tests Kits for Ammonium
Supplier: MACHEREY-NAGEL
Nitrite VHR L, 500 ml
Supplier: Lovibond
Sodium hydroxide 98%, flakes, pure
Supplier: Thermo Fisher Scientific