REAGENT KIT FOR PRIMELAB ID124(108TESTS) 1 * 1 items
Catalog # WTIDPL124-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID124(108TESTS) 1 * 1 items
Catalog # WTIDPL124-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
NANOCOLOR® Photometric Tube Tests, Nitrate
Supplier: MACHEREY-NAGEL
Sulphide reagents 1 and 2
Supplier: Hach
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond
Reagents for determination of silica
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach
BOD seed inoculum, POLYSEED
Supplier: Hach