25 TUBETESTS COD MR (0 - 1500 MG/L) 1 * 25 Vial
Catalog # WTIDCOD-80-MR
Supplier: WATER ID PRIMELAB
Frequently Bought Together
HEMATOGNOST Fe® stain for histology, Sigma-Aldrich®
Supplier: Merck
Accessories for NOVA and Picco
Supplier: Merck
Visual colour kits, zinc, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 35% in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Total Iron Test Strips, Dosatest, VWR®
Supplier: VWR Chemicals
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
pH indicator papers, universal/special
Supplier: MACHEREY-NAGEL
NANOCOLOR® Photometric Tube Tests, Nitrate
Supplier: MACHEREY-NAGEL
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Supplier: Honeywell Chemicals
NEISSER´S SOLUTION III FOR MICROSCOPY 1 * 100 mL
Supplier: Roth Carl
Test cassettes for urine
Supplier: SURESCREEN