Order Entry
Denmark
ContactUsLinkComponent
ZAG 130 dsDNA Separation Gel, 500mL. For use with the ZAG DNA Analyzer system and the ZAG-130 and ZAG-135 kits 1 * 500 mL
Catalog # AGLSZAG-130-0500
Supplier:  AGILENT
ZAG 130 dsDNA Separation Gel, 500mL. For use with the ZAG DNA Analyzer system and the ZAG-130 and ZAG-135 kits 1 * 500 mL
Catalog # AGLSZAG-130-0500
Supplier:  AGILENT
Frequently Bought Together