Order Entry
Denmark
ContactUsLinkComponent
Markers, 75 bp & 15 kb, 3.2 mL. 75 bp lower marker 15,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-9 20 kit. 1 * 3,2 mL
Catalog # AGLSFS-SMK920-0003
Supplier:  AGILENT
Markers, 75 bp & 15 kb, 3.2 mL. 75 bp lower marker 15,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-9 20 kit. 1 * 3,2 mL
Catalog # AGLSFS-SMK920-0003
Supplier:  AGILENT
Frequently Bought Together