0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
1 kb DNA Ladder, 100 µL. 250-10,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-920 kit. 1 * 0,1 mL
AGLSFS-SLR920-U100
:
AGILENT
1 kb DNA Ladder, 100 µL. 250-10,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-920 kit. 1 * 0,1 mL
AGLSFS-SLR920-U100
:
AGILENT
Frequently Bought Together
VWR® Blender Bags
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
Transfer pipettes
: RATIOLAB
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Disposable Microcentrifuge Tubes
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection