Dilution Buffer 0.1x TE, 60mL. Buffer for dilution of sample 1 * 60 mL
Catalog # AGLSDNF-494-0060
Supplier: AGILENT
Dilution Buffer 0.1x TE, 60mL. Buffer for dilution of sample 1 * 60 mL
Catalog # AGLSDNF-494-0060
Supplier: AGILENT
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Racks, Microcentrifuge Tube
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection