Order Entry
Denmark
ContactUsLinkComponent
5x Conditioning Solution, 50mL. Solution necessary for conditioning and restoring capillary array performance. 5x concentration. For use with the Femto Pulse system. 1 * 1 items
Catalog # AGLSDNF-425-0050
Supplier:  AGILENT
5x Conditioning Solution, 50mL. Solution necessary for conditioning and restoring capillary array performance. 5x concentration. For use with the Femto Pulse system. 1 * 1 items
Catalog # AGLSDNF-425-0050
Supplier:  AGILENT
Frequently Bought Together