Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hexamethyldisiloxane CAS:107-46-0 EC:203-492-7 1 * 1 mL
Catalog # CMPLSB25380.1000
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
Hexamethyldisiloxane CAS:107-46-0 EC:203-492-7 1 * 1 mL
Catalog # CMPLSB25380.1000
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
7 components: 1000mg/l each of As Ba Be Se Te U V in HNO3 10% ,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after opening),, Ambient Temp.,, 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
SULFITES 10 MG/L IN H2O 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
CHEMSET KIT TYPE K-7599 AMPULLEN 1 * 1 KIT
Supplier: CHEMetrics
Powder pillows, sulphate, Sulfa 4
Supplier: Lovibond
Reagents for DEHA determination with Lovibond® MD photometers
Supplier: Lovibond
KIT CHEMETS OXYGEN DISSOLVED 5-180 PPB 1 * 1 Each
Supplier: CHEMetrics
Cuvette tests for water testing for WTW spectrophotometers and photometers
Supplier: WTW