7 components: 1000mg/l each of As Ba Be Se Te U V in HNO3 10% ,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after opening),, Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM065085L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together