PYRROLIDINE-3-CARBOXYLIC ACID HYDROCHLOR 1 * 25 g
Catalog # APOSOR309145-25G
Supplier: Apollo Scientific
PYRROLIDINE-3-CARBOXYLIC ACID HYDROCHLOR 1 * 25 g
Catalog # APOSOR309145-25G
Supplier: Apollo Scientific
Frequently Bought Together
Acetone, anhydrous (max. 0.01% H₂O) ≥99.8%
Supplier: VWR Chemicals
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Acetone, J.T.Baker®
Supplier: Avantor
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Formaldehyde solution 35% (w/w) stabilised, meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99%
Supplier: Thermo Fisher Scientific