4-(2-PYRIDYL)ANILINE 1 * 25 g
APOSOR323239-25G
Frequently Bought Together
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
: VWR Chemicals