METHYL 3-CHLORO-4-IODOBENZOATE 1 * 5 g
APOSOR400970-5G
Frequently Bought Together
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
: Honeywell Chemicals
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Toluene ≥99.3%, Laboratory Reagent
: Honeywell Chemicals
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.7%, SPECTRONORM® for spectroscopy
: VWR Chemicals
Diethyl ether 99+% stabilised, pure
: Thermo Fisher Scientific
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
Formamide ≥99%
: Thermo Fisher Scientific
n-Pentane ≥99.0%, SPECTRONORM® for spectroscopy
: VWR Chemicals