2-Bromophenetole
Bottle, 1 g
Catalog # APOSOR9597-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Bromo-2-ethoxybenzene, 2-Bromophenyl ethyl ether
- CAS Number:583-19-7
- Formula:C₈H₉BrO
- MDL Number:MFCD00061092
- Molecular Weight:201.06 g/mol
- Boiling Pt:223 °C
- Density:1.4223 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:209-501-0
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



