Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Bromophenetole

2-Bromophenetole

Bottle, 100 mg
Catalog # APOSOR9597-100MG
Irritant
Supplier:   Apollo Scientific
2-Bromophenetole
Catalog # APOSOR9597-100MG
Supplier:  Apollo Scientific
CAS Number:  583-19-7
2-Bromophenetole

Bottle, 100 mg

CAS Number:  583-19-7
Price
165,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  1-Bromo-2-ethoxybenzene, 2-Bromophenyl ethyl ether
  • CAS Number:583-19-7
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00061092
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:223 °C
  • Density:1.4223 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:209-501-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg

Specifications