5-[(2-AMINOETHYL)AMINO]NAPHTHALENE-1-SUL 1 * 1 g
Catalog # APOSOR0920T-1G
Supplier: Apollo Scientific
5-[(2-AMINOETHYL)AMINO]NAPHTHALENE-1-SUL 1 * 1 g
Catalog # APOSOR0920T-1G
Supplier: Apollo Scientific
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Pyrogallol for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Acetone, meets analytical specification of Ph.Eur.
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
n-Hexane
Supplier: Roth Carl
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific