2-(3,5-DIFLUOROPHENYL)ETHANOL 1 * 5 g
Catalog # APOSPC400700-5G
Supplier: Apollo Scientific
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Butyl acetate ≥99.5%, Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Bromine 99.6% for analysis
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide 99.5% for analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
1-(Trimethylsilyl)imidazole, Supelco®
Supplier: Merck
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific