Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(+)-Biotin 99%

D-(+)-Biotin 99%

Bottle, 5 g
Catalog # APOSBIB0603-5G
Irritant
Supplier:   Apollo Scientific
D-(+)-Biotin 99%
Catalog # APOSBIB0603-5G
Supplier:  Apollo Scientific
CAS Number:  58-85-5
D-(+)-Biotin 99%

Bottle, 5 g

CAS Number:  58-85-5
Price
527,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Melting Pt:231…233 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications