CIS-2-PENTEN-1-OL 1 * 5 g
Catalog # APOSOR40816-5G
Supplier: Apollo Scientific
Frequently Bought Together
Xylen ≥98%, TECHNICAL
Supplier: VWR Chemicals
Toluen ≥98%, TECHNICAL
Supplier: VWR Chemicals
n-Pentan ≥95%, TECHNICAL
Supplier: VWR Chemicals
Cyclohexanon ≥99.0% analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dimethylsulfoxid, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dimethylsulfoxid, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Ethylacetat ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylacetat, Prepsolv® til præparativ kromatografi, Supelco®
Supplier: Merck
Toluen ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Heptan, TECHNICAL
Supplier: VWR Chemicals
Dichlormethan ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals