Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Cleanroom Socks

White
  ALSO1027438
Cleanroom Socks
  ALSO1027438
Cleanroom Socks

White

 
0,60 kr
   
1 * 1 Par
 

Alsico cleanroom socks minimises the risk of particle contamination in your controlled environment.

  • Unisex
  • One size tube socks
  • 97% polyester, 3% elastane
  • 2 variants: Thick and thin

Thin: white socks with blue Alsico high tech logo, thin quality: 167 Denier/48 Filament, 100% polyester
Thick: white socks with red Alsico high tech logo, thick quality: 150 Denier/288 Filament, 100% polyester

 : Packing: per 20 pairs.

 
  • Colour:
    White
  • Seq:
    01
  • Cat. No.:
    ALSO1027438
  • Model:
    Thick
  • Garment application:
    Cleanroom
  • Gender:
    Unisex
  • Fabric:
    97% polyester and 3% elastane
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.