Order Entry
Denmark
ContactUsLinkComponent
Leica M205 C optics carrier, with 20.5:1 encoded zoom - Magnification range: 7.8x-160x (1.0x objective, 10x eyepieces) - Coded functions: magnification and iris Diaphragm - Apochromatic corrected optics - For parfocal planapochromat objectives: 0.63 1 * 1 items
Catalog # LEIM10450036
Supplier:  LEICA MICROSYSTEMS
Leica M205 C optics carrier, with 20.5:1 encoded zoom - Magnification range: 7.8x-160x (1.0x objective, 10x eyepieces) - Coded functions: magnification and iris Diaphragm - Apochromatic corrected optics - For parfocal planapochromat objectives: 0.63 1 * 1 items
Catalog # LEIM10450036
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together