Order Entry
Denmark
ContactUsLinkComponent
Sontara Micropure AP White 305x305mm, cleanroom & electronics nonwoven wipe (Recommended for ISO 6-8* cleanrooms): Strong, durable, absorbent, low-linting, 20 packs x 150 wipes per box. 1 * 3.000 items
Catalog # SONTD13650646
Supplier:  SONTARA
Sontara Micropure AP White 305x305mm, cleanroom & electronics nonwoven wipe (Recommended for ISO 6-8* cleanrooms): Strong, durable, absorbent, low-linting, 20 packs x 150 wipes per box. 1 * 3.000 items
Catalog # SONTD13650646
Supplier:  SONTARA
Frequently Bought Together