Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

D-(+)-Glucose monohydrate, powder, TECHNICAL

Plastic drum, 25 kg
Catalog # 24374.460
Supplier:   VWR Chemicals
D-(+)-Glucose monohydrate, powder, TECHNICAL
Catalog # 24374.460
Supplier:  VWR Chemicals
CAS Number:  14431-43-7
D-(+)-Glucose monohydrate, powder, TECHNICAL
D-(+)-Glucose monohydrate, powder, TECHNICAL

Plastic drum, 25 kg

CAS Number:  14431-43-7
Price
3.960,00 kr
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
  • CAS Number:14431-43-7
  • Formula:C₆H₁₂O₆·H₂O
  • MDL Number:MFCD00149450
  • Molecular Weight:198.17 g/mol
  • Melting Pt:83 °C
  • Storage Temperature:Ambient
  • EINECS:200-075-1
Specifications

Specification Test Results

  • Water:
    7.0 - 9.5 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    25 kg

Specifications