Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dextrin, white powder

Dextrin, white powder

Poly bottle, 500 g
Catalog # SPCMDE137-500GM
Supplier:   Spectrum Chemical
Dextrin, white powder
Catalog # SPCMDE137-500GM
Supplier:  Spectrum Chemical
CAS Number:  9004-53-9
Dextrin, white powder

Poly bottle, 500 g

CAS Number:  9004-53-9
Price
763,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Dextrin, White Powder is used as water-soluble glue in envelope adhesives, crispness enhancers in food processing and as a thickening agent in pharmaceuticals.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Storage Temperature:Ambient
  • EINECS:232-675-4
Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.