Dextrin, white powder
Poly bottle, 2,5 kg
Catalog # SPCMDE137-2.5KG
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Dextrin, White Powder is used as water-soluble glue in envelope adhesives, crispness enhancers in food processing and as a thickening agent in pharmaceuticals.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Synonyms: Dextrin
- CAS Number:9004-53-9
- Formula:(C₆H₁₂O₆)n
- MDL Number:MFCD00081554
- Molecular Weight:180.16 g/mol
- Storage Temperature:Ambient
- EINECS:232-675-4
Specifications
Specifications
- Pack type:Poly bottle
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



