0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
[EN]PYJAMAS MEDIAL BLUE XXLARGE 10X10PCS 1 * 100 Items
KING520216/10
:
KING DISPOSABLES
[EN]PYJAMAS MEDIAL BLUE XXLARGE 10X10PCS 1 * 100 Items
KING520216/10
:
KING DISPOSABLES
Frequently Bought Together
Handsker, nitril, Kimtech™ Purple Nitrile™ Xtra™
: KIMBERLY CLARK
Tape, temperaturresistent
: PRECISION DYNAMICS TIMEM
VWR® Nitrile Light Long, Nitrile Gloves
: VWR Collection
Handsker, nitril, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Cleanroom gloves, polychloroprene, sterile, BioClean Ultimate™
: Nitritex
GHS-piktogrammer
: Brady
VWR® Nitrile Extra Light, Nitrile Gloves
: VWR Collection
VWR®, Sikkerhedsmærkater i henhold til GHS
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Latex Gloves
: VWR Collection
VWR® Nitrile Green Nitrile gloves
: VWR Collection