TOTAL HARDNESS 1000 MG/L CACO3 STANDARD 1 * 100 mL
Catalog # ACSDHRDN1000.L1
Supplier: CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW