Order Entry
Denmark
ContactUsLinkComponent
Volumetric flask, 5 ml, class A, conformity certified, ST7/16, ST-hollow glass stopper, Diffico brown, made from DURAN tubing 1 * 10 items
Catalog # WITG3.668.000
Supplier:  witeg Labortechnik
Volumetric flask, 5 ml, class A, conformity certified, ST7/16, ST-hollow glass stopper, Diffico brown, made from DURAN tubing 1 * 10 items
Catalog # WITG3.668.000
Supplier:  witeg Labortechnik
Frequently Bought Together