MYXAL SEPT GEL 1L HARTFLASCHE 1 * 10 items
Catalog # FAWE13743-003
Supplier: FAWECO HAUTSCHUTZ
Frequently Bought Together
Methanol anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Water for analysis
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C ACS
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Pyridine 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
p-Xylene ≥99.0% (by GC)
Supplier: TCI
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific