TESTPAK CHECKIT COMPARATOR CHLORINE DPD 1 * 1 KIT
Catalog # LOVI147540
Supplier: Lovibond
Frequently Bought Together
VWR® Q Path® DC3, Decalcifier Rapid-Acting
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Visual colour kits, iron, MQuant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
Test kits, photometric, Nitrite, LCK cuvette tests
Supplier: Hach
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Test kits, photometric, iron, LCK cuvette tests
Supplier: Hach