Order Entry
Denmark
ContactUsLinkComponent
BD Vacutainer® Tubes for Trace Element Determination, Plastic 6,0 ml, 13 x 100 mm, K2 EDTA, Royal Blue, Hemogard™ Closure Paper Packing 10x100 pcs 1 * 100 items
Catalog # BDAM368381
Supplier:  BD Medical
BD Vacutainer® Tubes for Trace Element Determination, Plastic 6,0 ml, 13 x 100 mm, K2 EDTA, Royal Blue, Hemogard™ Closure Paper Packing 10x100 pcs 1 * 100 items
Catalog # BDAM368381
Supplier:  BD Medical
Frequently Bought Together