Order Entry
Denmark
ContactUsLinkComponent
Autoclave 3870ELVCPVG-380-D, Orientation : Vertical, Volume : 85 L, Options: Fast cooling (up to 75%) / Complete drying (steam generator and vacuum). 1 * 1 items
Catalog # TUTT3870ELVCPVG380
Supplier:  TUTTNAUER
Autoclave 3870ELVCPVG-380-D, Orientation : Vertical, Volume : 85 L, Options: Fast cooling (up to 75%) / Complete drying (steam generator and vacuum). 1 * 1 items
Catalog # TUTT3870ELVCPVG380
Supplier:  TUTTNAUER
Frequently Bought Together