SILICONE (ANTI-FOAMING AGENT) 1 * 500 g
Catalog # HIMEMB158-500G
Supplier: HIMEDIA
Frequently Bought Together
Benzyl alcohol, EMSURE® for analysis, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥99.0% (by GC), LiChrosolv®, hypergrade for LC-MS
Supplier: Merck
Methyl ethyl ketone, EMPLURA®, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
n-Hexane ≥99.0%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Supplier: Merck
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals