Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

CryoStor® CSB Cryopreservation Medium

CryoStor® CSB, 500 ml, Human; Animal component-free, chemically defined, serum-free, 1 KIT
Catalog # STMC100-0238
Marketsource
CryoStor® CSB Cryopreservation Medium
Catalog # STMC100-0238
Marketsource
CAS Number:  
CryoStor® CSB Cryopreservation Medium
CryoStor® CSB Cryopreservation Medium

CryoStor® CSB, 500 ml, Human; Animal component-free, chemically defined, serum-free, 1 KIT

CAS Number:  
Price
28.054,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Animal component-free and protein-free cryopreservation formulation.

  • Serum-free and chemically defined
  • USP grade/multi-compendial/high-quality components
  • cGMP-manufactured
  • Sterility, endotoxin, and cell-based release testing
  • FDA master file

Reduce the level of cryopreservation-induced delayed-onset cell death and maximise post-thaw cell recovery, viability, and function following cryopreservation, with CryoStor® CSB, an intracellular-like base formulation from the CryoStor® preservation media series. Serum-free, animal component-free, and cGMP-manufactured, this chemically defined medium is sterile and tested for endotoxins and cell-based release. CryoStor® CSB is formulated with USP-grade components and does not contain DMSO.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    CryoStor® CSB, 500 ml
  • Format:
    Human; Animal component-free, chemically defined, serum-free

Specifications