Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant BMP-4

Human Recombinant BMP-4

100 µG
Catalog # STMC78211.1
Marketsource
Human Recombinant BMP-4
Catalog # STMC78211.1
Marketsource
CAS Number:  
Human Recombinant BMP-4
Human Recombinant BMP-4

100 µG

CAS Number:  
Price
10.459,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Bone morphogenetic protein 4 (BMP-4) is a member of the highly conserved transforming growth factor β (TGF-β) superfamily. Mature BMP-4 is a disulfide-linked homodimeric protein consisting of two 116-amino-acid residue subunits, and is generated by the proteolytic removal of the signal peptide and propeptides (Xiao et al.). BMP-4 binds to type I and type II receptors on cells. This binding results in the phosphorylation of receptor 1, which in turn results in the phosphorylation of Smad proteins, which then go on to act as transcription factors (Zhang et al.). BMPs have been shown to be key regulators of embryogenesis and are known to play a role in the growth and differentiation of various cell types, including embryonic stem (ES) cells, induced pluripotent stem (iPS) cells, mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells (Chadwick et al.; Graham et al.; Jones et al.; Lengerke et al.; Zhang et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    STMC78211.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    BMP-4
  • Purity:
    ≥95%
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.