Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Human/Mouse Recombinant TGF-beta 3, ACF

10 µG
  STMC78156
Marketsource
Human/Mouse Recombinant TGF-beta 3, ACF
  STMC78156
Marketsource
 :  
Human/Mouse Recombinant TGF-beta 3, ACF
Human/Mouse Recombinant TGF-beta 3, ACF

10 µG

 :  
 
1.791,00 kr
   
1 * 1 Each
 

Transforming growth factor (TGF) beta 3 is a member of the TGF-β superfamily and regulates diverse cellular phenotypes. TGF-beta 3 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins, thus regulating a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (Massagué; McDowell et al.). TGF-beta 3 enhances glycosaminoglycan production by mesenchymal stromal cells, stimulates scar-free healing, and improves glucose tolerance and phenotypic changes in adipocyte morphology (Hall et al.; Holton et al.). TGF-beta 3 induces proliferation of posterofrontal suture-derived mesenchymal stromal cells, and stimulates expression of fibroblast growth factors 2 and 18 (James et al.). This product is animal component-free.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

 

 

  • Pk:
    10 µG
  • Cat. No.:
    STMC78156
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    TGF-beta 3, ACF
  • Purity:
    ≥95%
  • Species:
    Human|Mouse

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.