REAG IRON AMPOULES 1 * 1 KIT
Catalog # CHETR-6201
Supplier: CHEMetrics
Frequently Bought Together
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Mounting medium, Entellan® new
Supplier: Merck
Lugol's iodine solution, Sigma-Aldrich®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, manganese, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
VWR®, OCT Embedding matrix for frozen sections
Supplier: VWR Chemicals
Standards, calibration for LCK cuvettes, Addista
Supplier: Hach
Test kits, photometric, iron, LCK cuvette tests
Supplier: Hach
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach